Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044605.1 | NP_001038070.1 | 1 - 2 | 235 | 86 | |
XM_021066261.1 | XP_020921920.1 | 867 | 86 | |||
GenBank mRNA | DQ323981.1 | - | 1 - 2 | 235 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |