Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044605.1 | NP_001038070.1 | 3 - 4 | 453 | 72 | |
GenBank mRNA | DQ323981.1 | - | 3 - 4 | 453 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |