Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077214.1 | NP_001070682.1 | 2 - 3 | 260 | 54 | |
XM_021072205.1 | XP_020927864.1 | 325 | 54 | |||
XM_021072206.1 | XP_020927865.1 | 393 | 54 | |||
GenBank mRNA | DQ923315.1 | - | 2 - 3 | 260 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |