Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101033.1 | NP_001094503.1 | 14 - 15 | 1423 | 74 | |
XM_005672829.3 | XP_005672886.1 | 2922 | 74 | |||
GenBank mRNA | AK237864.1 | - | 15 - 16 | 1429 | 74 | |
AK350647.1 | - | 14 - 15 | 1440 | 74 | ||
DQ859019.1 | - | 14 - 15 | 1423 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |