Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101825.1 | NP_001095295.1 | 10 - 11 | 1129 | 63 | |
XM_005665804.3 | XP_005665861.1 | 1105 | 63 | |||
GenBank mRNA | AK233579.1 | - | 10 - 11 | 1114 | 63 | |
AK236334.1 | - | 10 - 11 | 1253 | 63 | ||
AK389553.1 | - | 10 - 11 | 1139 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |