Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110174.1 | NP_001103644.1 | 6 - 7 | 1329 | 63 | |
XM_013985484.2 | XP_013840938.1 | 1344 | 63 | |||
XM_013985485.2 | XP_013840939.1 | 1344 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |