Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113217.1 | NP_001106688.1 | 6 - 7 | 794 | 95 | |
GenBank mRNA | AK343747.1 | - | 6 - 7 | 779 | 95 | |
AK343823.1 | - | 6 - 7 | 770 | 95 | ||
AY550040.1 | - | 3 - 4 | 398 | 95 | ||
AY735454.1 | - | 6 - 7 | 794 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |