Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113704.1 | NP_001107176.1 | 5 - 6 | 1112 | 71 | |
GenBank mRNA | AF464006.1 | - | 5 - 6 | 1112 | 71 | |
AF464007.1 | - | 5 - 6 | 1112 | 71 | ||
AF464008.1 | - | 5 - 6 | 1112 | 71 | ||
AF464019.1 | - | 5 - 6 | 1112 | 71 | ||
AF464020.2 | - | 5 - 6 | 1074 | 71 | ||
AK236916.1 | - | 4 - 5 | 832 | 71 | ||
AK239694.1 | - | 5 - 6 | 1093 | 71 | ||
AK348712.1 | - | 4 - 5 | 821 | 71 | ||
AK392987.1 | - | 5 - 6 | 1097 | 71 | ||
AK394974.1 | - | 4 - 5 | 821 | 71 | ||
AY247771.1 | - | 5 - 6 | 1112 | 71 | ||
AY247772.1 | - | 5 - 6 | 1112 | 71 | ||
AY459304.1 | - | 5 - 6 | 1112 | 71 | ||
AY463538.1 | - | 2 - 3 | 231 | 71 | ||
AY463540.1 | - | 5 - 6 | 1072 | 71 | ||
DQ883214.1 | - | 5 - 6 | 1110 | 71 | ||
DQ883215.1 | - | 5 - 6 | 1112 | 71 | ||
DQ883216.1 | - | 5 - 6 | 1112 | 71 | ||
DQ976363.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992502.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992503.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992504.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992505.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992506.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992507.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992508.1 | - | 5 - 6 | 1100 | 71 | ||
DQ992509.1 | - | 5 - 6 | 1100 | 71 | ||
EU432054.1 | - | 5 - 6 | 1093 | 71 | ||
EU432055.1 | - | 5 - 6 | 1093 | 71 | ||
EU432056.1 | - | 5 - 6 | 1093 | 71 | ||
EU432057.1 | - | 5 - 6 | 1093 | 71 | ||
GU322911.1 | - | 5 - 6 | 1078 | 71 | ||
GU322912.1 | - | 4 - 5 | 802 | 71 | ||
GU322915.1 | - | 4 - 5 | 1669 | 71 | ||
GU322916.1 | - | 2 - 3 | 177 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |