Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123082.1 | NP_001116554.1 | 2 - 3 | 273 | 66 | |
XM_021077205.1 | XP_020932864.1 | 313 | 66 | |||
GenBank mRNA | AK392477.1 | - | 2 - 3 | 210 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |