Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123143.1 | NP_001116615.1 | 3 - 4 | 368 | 121 | |
XM_005664831.3 | XP_005664888.1 | 1051 | 121 | |||
GenBank mRNA | AB516285.1 | - | 3 - 4 | 398 | 121 | |
AK396179.1 | - | 3 - 4 | 397 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |