Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123160.1 | NP_001116632.1 | 3 - 4 | 487 | 135 | |
GenBank mRNA | AK391681.1 | - | 3 - 4 | 280 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |