Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213935.2 | NP_999100.2 | 1 - 2 | 283 | 69 | |
GenBank mRNA | AF059493.1 | - | 1 - 2 | 108 | 69 | |
AK237007.1 | - | 1 - 2 | 203 | 69 | ||
DQ917650.1 | - | 1 - 2 | 150 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |