Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213983.1 | NP_999148.1 | 2 - 3 | 243 | 69 | |
XM_013980839.2 | XP_013836293.1 | 375 | 69 | |||
XM_021066210.1 | XP_020921869.1 | 192 | 69 | |||
XM_021066211.1 | XP_020921870.1 | 203 | 69 | |||
GenBank mRNA | AK234194.1 | - | 2 - 3 | 133 | 69 | |
AK395258.1 | - | 2 - 3 | 155 | 69 | ||
AK396074.1 | - | 2 - 3 | 119 | 69 | ||
X89719.1 | - | 71 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Inventoried Probe spans exons |