Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213995.1 | NP_999160.1 | 8 - 9 | 1434 | 58 | |
GenBank mRNA | AB018743.1 | - | 8 - 9 | 1434 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |