Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214008.1 | NP_999173.1 | 4 - 5 | 585 | 62 | |
GenBank mRNA | AY077683.1 | - | 4 - 5 | 585 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |