Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001265.1 | NP_001001265.1 | 3 - 4 | 224 | 64 | |
GenBank mRNA | AF148221.1 | - | 3 - 4 | 224 | 64 | |
AK230637.1 | - | 3 - 4 | 253 | 64 | ||
AK230931.1 | - | 3 - 4 | 273 | 64 | ||
AK231016.1 | - | 3 - 4 | 273 | 64 | ||
AK231062.1 | - | 3 - 4 | 300 | 64 | ||
AK231092.1 | - | 3 - 4 | 266 | 64 | ||
AK231129.1 | - | 3 - 4 | 247 | 64 | ||
AK343716.1 | - | 3 - 4 | 306 | 64 | ||
AK343723.1 | - | 3 - 4 | 247 | 64 | ||
AK343862.1 | - | 3 - 4 | 247 | 64 | ||
AK343884.1 | - | 3 - 4 | 247 | 64 | ||
AK343896.1 | - | 3 - 4 | 247 | 64 | ||
AK343918.1 | - | 3 - 4 | 266 | 64 | ||
AK343960.1 | - | 3 - 4 | 247 | 64 | ||
AK343964.1 | - | 3 - 4 | 266 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |