Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001770.1 | NP_001001770.1 | 3 - 4 | 363 | 105 | |
GenBank mRNA | AF016388.1 | - | 3 - 4 | 363 | 105 | |
AK236522.1 | - | 3 - 4 | 415 | 105 | ||
AY609822.1 | - | 3 - 4 | 393 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |