Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005153.1 | NP_001005153.1 | 2 - 3 | 244 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |