Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033013.2 | NP_001028185.1 | 5 - 6 | 860 | 106 | |
XM_005663159.3 | XP_005663216.1 | 868 | 106 | |||
XM_013996819.2 | XP_013852273.1 | 925 | 106 | |||
GenBank mRNA | AK239470.1 | - | 6 - 7 | 913 | 106 | |
AK391270.1 | - | 5 - 6 | 865 | 106 | ||
DQ026064.1 | - | 5 - 6 | 863 | 106 | ||
DQ027013.1 | - | 5 - 6 | 863 | 106 | ||
HM122239.1 | - | 4 - 5 | 536 | 106 | ||
HM803103.1 | - | 5 - 6 | 788 | 106 | ||
HM803104.1 | - | 5 - 6 | 791 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |