Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044528.2 | NP_001037993.1 | 4 - 5 | 369 | 72 | |
XM_005666483.3 | XP_005666540.1 | 770 | 72 | |||
XM_021100516.1 | XP_020956175.1 | 1631 | 72 | |||
GenBank mRNA | AK234815.1 | - | 6 - 7 | 644 | 72 | |
EU671074.1 | - | 4 - 5 | 369 | 72 | ||
KU705638.1 | - | 4 - 5 | 369 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |