Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213768.1 | NP_998933.1 | 5 - 6 | 992 | 74 | |
XM_005665788.3 | XP_005665845.2 | 1167 | 74 | |||
XM_013977626.2 | XP_013833080.2 | 1137 | 74 | |||
XM_021098282.1 | XP_020953941.1 | 1450 | 74 | |||
GenBank mRNA | AY463541.1 | - | 5 - 6 | 992 | 74 | |
GU322918.1 | - | 5 - 6 | 992 | 74 | ||
GU322919.1 | - | 2 - 3 | 177 | 74 | ||
GU322920.1 | - | 2 - 3 | 177 | 74 | ||
HQ224544.1 | - | 6 - 7 | 893 | 74 | ||
HQ224545.1 | - | 6 - 7 | 970 | 74 | ||
HQ224547.1 | - | 5 - 6 | 970 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |