Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213882.1 | NP_999047.1 | 1 - 2 | 140 | 114 | |
GenBank mRNA | AK348522.1 | - | 1 - 2 | 400 | 114 | |
X07383.1 | - | 140 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |