Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213960.1 | NP_999125.1 | 7 | 746 | 75 | |
XM_005652948.3 | XP_005653005.1 | 875 | 75 | |||
XM_005652949.3 | XP_005653006.1 | 853 | 75 | |||
XM_005652950.3 | XP_005653007.1 | 913 | 75 | |||
XM_021080765.1 | XP_020936424.1 | 894 | 75 | |||
GenBank mRNA | AF268461.1 | - | 7 | 746 | 75 | |
AK235603.1 | - | 8 | 817 | 75 | ||
AK391290.1 | - | 8 | 828 | 75 | ||
AK392730.1 | - | 8 | 789 | 75 | ||
AK393361.1 | - | 8 | 817 | 75 | ||
AK393578.1 | - | 8 | 801 | 75 | ||
AK395118.1 | - | 8 | 821 | 75 | ||
AK400488.1 | - | 8 | 886 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Inventoried Amplicon spans exons and probe does not span exons |