Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213990.1 | NP_999155.1 | 5 - 6 | 445 | 76 | |
GenBank mRNA | AF285449.1 | - | 4 - 5 | 385 | 76 | |
AF285450.1 | - | 5 - 6 | 445 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |