Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214123.1 | NP_999288.1 | 3 - 4 | 335 | 67 | |
GenBank mRNA | AF493991.1 | - | 3 - 4 | 318 | 67 | |
AY294020.1 | - | 3 - 4 | 324 | 67 | ||
HQ236500.1 | - | 3 - 4 | 312 | 67 | ||
JF906512.1 | - | 3 - 4 | 312 | 67 | ||
L12991.1 | - | 3 - 4 | 358 | 67 | ||
X68330.1 | - | 335 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |