Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214200.2 | NP_999365.1 | 6 - 7 | 855 | 67 | |
XM_005660074.3 | XP_005660131.1 | 892 | 67 | |||
XM_005660075.3 | XP_005660132.1 | 1035 | 67 | |||
XM_013993524.2 | XP_013848978.1 | 946 | 67 | |||
XM_021082975.1 | XP_020938634.1 | 891 | 67 | |||
GenBank mRNA | AB527057.1 | - | 5 - 6 | 780 | 67 | |
AF234676.2 | - | 6 - 7 | 856 | 67 | ||
AK232656.1 | - | 6 - 7 | 855 | 67 | ||
AY550037.1 | - | 6 - 7 | 853 | 67 | ||
DQ066659.1 | - | 5 - 6 | 780 | 67 | ||
JN616387.1 | - | 5 - 6 | 780 | 67 | ||
KC733245.1 | - | 5 - 6 | 780 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |