Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214279.1 | NP_999444.1 | 3 - 4 | 211 | 139 | |
GenBank mRNA | AF527991.1 | - | 3 - 4 | 211 | 139 | |
AK232253.1 | - | 3 - 4 | 214 | 139 | ||
AK234031.1 | - | 3 - 4 | 206 | 139 | ||
AK346956.1 | - | 3 - 4 | 214 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |