Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214344.1 | NP_999509.1 | 4 - 5 | 560 | 101 | |
XM_021098311.1 | XP_020953970.1 | 627 | 101 | |||
XM_021098312.1 | XP_020953971.1 | 627 | 101 | |||
XM_021098313.1 | XP_020953972.1 | 591 | 101 | |||
XM_021098314.1 | XP_020953973.1 | 627 | 101 | |||
XM_021098315.1 | XP_020953974.1 | 626 | 101 | |||
GenBank mRNA | AB104465.1 | - | 4 - 5 | 560 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |