Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214364.1 | NP_999529.1 | 4 - 5 | 319 | 86 | |
XM_021077307.1 | XP_020932966.1 | 473 | 86 | |||
XM_021077308.1 | XP_020932967.1 | 542 | 86 | |||
GenBank mRNA | AF268463.1 | - | 4 - 5 | 319 | 86 | |
AK344428.1 | - | 5 - 6 | 376 | 86 | ||
JX280457.1 | - | 4 - 5 | 286 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |