Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC106506651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013985231.2 | XP_013840685.1 | 336 | 101 | ||
GenBank mRNA | AK397272.1 | - | 1 | 324 | 101 | |
AK397426.1 | - | 1 | 324 | 101 | ||
AM157714.1 | - | 1 | 252 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |