Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141991.2 | NP_001135463.2 | 3 | 465 | 77 | |
GenBank mRNA | AK240144.1 | - | 3 | 447 | 77 | |
AK392004.1 | - | 3 | 449 | 77 | ||
FJ234413.1 | - | 3 | 452 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |