Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143711.1 | NP_001137183.1 | 4 | 546 | 64 | |
GenBank mRNA | FJ439674.1 | - | 4 | 546 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |