Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164514.1 | NP_001157986.1 | 2 - 3 | 136 | 100 | |
XM_021066050.1 | XP_020921709.1 | 192 | 100 | |||
XM_021066051.1 | XP_020921710.1 | 192 | 100 | |||
GenBank mRNA | GQ368240.1 | - | 2 - 3 | 136 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |