Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167643.1 | NP_001161115.1 | 4 - 5 | 575 | 70 | |
GenBank mRNA | AB529450.1 | - | 4 - 5 | 575 | 70 | |
AK235587.1 | - | 4 - 5 | 839 | 70 | ||
AK395698.1 | - | 5 - 6 | 692 | 70 | ||
AY609406.1 | - | 4 - 5 | 805 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |