Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128472.1 | NP_001121944.1 | 1 - 2 | 96 | 76 | |
GenBank mRNA | AK347456.1 | - | 1 - 2 | 129 | 76 | |
AK347693.1 | - | 1 - 2 | 125 | 76 | ||
EF581989.1 | - | 1 - 2 | 96 | 76 | ||
EU429459.1 | - | 1 - 2 | 1024 | 76 | ||
EU429460.1 | - | 1 - 2 | 756 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |