Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145384.1 | NP_001138856.1 | 2 - 3 | 408 | 84 | |
GenBank mRNA | JN966758.1 | - | 2 - 3 | 578 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Inventoried Probe spans exons |