Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159615.1 | NP_001153087.1 | 4 - 5 | 777 | 81 | |
GenBank mRNA | AK350973.1 | - | 4 - 5 | 782 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |