Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160081.1 | NP_001153553.1 | 4 - 5 | 755 | 102 | |
XM_021068338.1 | XP_020923997.1 | 668 | 102 | |||
GenBank mRNA | FJ875103.1 | - | 4 - 5 | 755 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |