Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170521.1 | NP_001163992.1 | 4 - 5 | 738 | 75 | |
XM_005660695.3 | XP_005660752.2 | 959 | 75 | |||
XM_021077409.1 | XP_020933068.1 | 769 | 75 | |||
GenBank mRNA | FJ446485.1 | - | 4 - 5 | 738 | 75 | |
KP717445.1 | - | 4 - 5 | 738 | 75 | ||
KP717446.1 | - | 5 - 6 | 671 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |