Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1LC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170827.1 | NP_001164298.1 | 2 - 3 | 248 | 73 | |
GenBank mRNA | AK240181.1 | - | 2 - 3 | 248 | 73 | |
AK347670.1 | - | 2 - 3 | 216 | 73 | ||
AK392137.1 | - | 2 - 3 | 218 | 73 | ||
AK392455.1 | - | 2 - 3 | 243 | 73 | ||
GU272221.1 | - | 2 - 3 | 248 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |