Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170828.1 | NP_001164299.1 | 4 - 5 | 684 | 62 | |
XM_005656037.3 | XP_005656094.3 | 799 | 62 | |||
GenBank mRNA | GU121641.1 | - | 4 - 5 | 684 | 62 | |
GU373697.1 | - | 4 - 5 | 589 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |