Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177922.1 | NP_001171393.1 | 2 - 3 | 206 | 86 | |
XM_013995173.2 | XP_013850627.1 | 241 | 86 | |||
GenBank mRNA | AK392880.1 | - | 2 - 3 | 207 | 86 | |
AK393793.1 | - | 2 - 3 | 209 | 86 | ||
GU475005.1 | - | 2 - 3 | 206 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |