Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185049.2 | NP_001171978.2 | 10 - 11 | 3085 | 63 | |
XM_021100320.1 | XP_020955979.1 | 3109 | 63 | |||
XM_021100322.1 | XP_020955981.1 | 3028 | 63 | |||
XM_021100323.1 | XP_020955982.1 | 3019 | 63 | |||
XM_021100324.1 | XP_020955983.1 | 3004 | 63 | |||
XM_021100325.1 | XP_020955984.1 | 2995 | 63 | |||
XM_021100326.1 | XP_020955985.1 | 1527 | 63 | |||
XM_021100327.1 | XP_020955986.1 | 4544 | 63 | |||
GenBank mRNA | HM037344.1 | - | 9 - 10 | 1150 | 63 | |
HM037345.1 | - | 9 - 10 | 1141 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |