Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185136.1 | NP_001172065.1 | 1 - 2 | 127 | 62 | |
GenBank mRNA | AB529448.1 | - | 1 - 2 | 127 | 62 | |
AK236423.1 | - | 1 - 2 | 147 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |