Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185151.1 | NP_001172080.1 | 2 - 3 | 111 | 80 | |
XM_005666270.3 | XP_005666327.1 | 125 | 80 | |||
XM_021098131.1 | XP_020953790.1 | 143 | 80 | |||
GenBank mRNA | AK238583.1 | - | 2 - 3 | 128 | 80 | |
AK392634.1 | - | 2 - 3 | 103 | 80 | ||
AY609459.1 | - | 2 - 3 | 96 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |