Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185171.1 | NP_001172100.1 | 7 - 8 | 801 | 97 | |
XM_005671491.3 | XP_005671548.1 | 637 | 97 | |||
XM_005671493.3 | XP_005671550.1 | 553 | 97 | |||
XM_005671494.3 | XP_005671551.1 | 582 | 97 | |||
XM_005671498.3 | XP_005671555.1 | 515 | 97 | |||
XM_021071868.1 | XP_020927527.1 | 725 | 97 | |||
GenBank mRNA | AK234444.1 | - | 7 - 8 | 799 | 96 | |
AK238763.1 | - | 5 - 6 | 511 | 97 | ||
AK351475.1 | - | 5 - 6 | 502 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |