Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194974.1 | NP_001181903.1 | 14 - 15 | 1839 | 60 | |
XM_021093288.1 | XP_020948947.1 | 2105 | 60 | |||
XM_021093289.1 | XP_020948948.1 | 2200 | 60 | |||
XM_021093290.1 | XP_020948949.1 | 1882 | 60 | |||
XM_021093291.1 | XP_020948950.1 | 2128 | 60 | |||
GenBank mRNA | GQ329716.1 | - | 14 - 15 | 1839 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |