Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194974.1 | NP_001181903.1 | 15 - 16 | 2036 | 98 | |
XM_021093288.1 | XP_020948947.1 | 2302 | 98 | |||
XM_021093289.1 | XP_020948948.1 | 2397 | 98 | |||
XM_021093290.1 | XP_020948949.1 | 2079 | 98 | |||
XM_021093291.1 | XP_020948950.1 | 2325 | 98 | |||
GenBank mRNA | GQ329716.1 | - | 15 - 16 | 2036 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |