Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195326.1 | NP_001182255.1 | 17 - 18 | 1485 | 109 | |
XM_021098060.1 | XP_020953719.1 | 3069 | 109 | |||
GenBank mRNA | AK392315.1 | - | 17 - 18 | 1479 | 109 | |
EU282343.1 | - | 17 - 18 | 1485 | 109 | ||
KX068708.1 | - | 17 - 18 | 1445 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |