Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195362.1 | NP_001182291.1 | 8 - 9 | 724 | 95 | |
XM_005657214.3 | XP_005657271.2 | 717 | 95 | |||
XM_005657216.3 | XP_005657273.2 | 1595 | 95 | |||
GenBank mRNA | GU373718.1 | - | 8 - 9 | 724 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |